EMAIL THIS PAGE TO A FRIEND
Journal of Thoracic Oncology : Official Publication of the International Association for the Study of Lung Cancer
Detection of human polyomavirus 7 in human thymic epithelial tumors.
PMID 25526237