EMAIL THIS PAGE TO A FRIEND
Parasites & vectors
Anopheles gambiae salivary protein expression modulated by wild Plasmodium falciparum infection: highlighting of new antigenic peptides as candidates of An. gambiae bites.
PMID 25526764