EMAIL THIS PAGE TO A FRIEND
BMC biology
A substrate ambiguous enzyme facilitates genome reduction in an intracellular symbiont.
PMID 25527092
Abstract
Genome evolution in intracellular microbial symbionts is characterized by gene loss, generating some of the smallest and most gene-poor genomes known. As a result of gene loss these genomes commonly contain metabolic pathways that are fragmented relative to their free-living relatives. The evolutionary retention of fragmented metabolic pathways in the gene-poor genomes of endosymbionts suggests that they are functional. However, it is not always clear how they maintain functionality. To date, the fragmented metabolic pathways of endosymbionts have been shown to maintain functionality through complementation by host genes, complementation by genes of another endosymbiont and complementation by genes in host genomes that have been horizontally acquired from a microbial source that is not the endosymbiont. Here, we demonstrate a fourth mechanism. We investigate the evolutionary retention of a fragmented pathway for the essential nutrient pantothenate (vitamin B5) in the pea aphid, Acyrthosiphon pisum endosymbiosis with Buchnera aphidicola. Using quantitative analysis of gene expression we present evidence for complementation of the Buchnera pantothenate biosynthesis pathway by host genes. Further, using complementation assays in an Escherichia coli mutant we demonstrate functional replacement of a pantothenate biosynthesis enzyme, 2-dehydropantoate 2-reductase (E.C. 1.1.1.169), by an endosymbiont gene, ilvC, encoding a substrate ambiguous enzyme. Earlier studies have speculated that missing enzyme steps in fragmented endosymbiont metabolic pathways are completed by adaptable endosymbiont enzymes from other pathways. Here, we experimentally demonstrate completion of a fragmented endosymbiont vitamin biosynthesis pathway by recruitment of a substrate ambiguous enzyme from another pathway. In addition, this work extends host/symbiont metabolic collaboration in the aphid/Buchnera symbiosis from amino acid metabolism to include vitamin biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8731
Calcium pantothenate, meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)
C9H16NO5 · 0.5Ca
C0400000
Calcium pantothenate, European Pharmacopoeia (EP) Reference Standard
C9H16NO5 · 0.5Ca
1087009
Calcium Pantothenate, United States Pharmacopeia (USP) Reference Standard
C9H16NO5 · 0.5Ca
P5155
D-Pantothenic acid hemicalcium salt, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C9H16NO5 · 0.5Ca
21210
D-Pantothenic acid hemicalcium salt, ≥98.0%
C9H16NO5 · 0.5Ca
47867
D-Pantothenic acid hemicalcium salt, analytical standard
C9H16NO5 · 0.5Ca
I0460000
Isoleucine, European Pharmacopoeia (EP) Reference Standard
C6H13NO2
W527602
L-Isoleucine, 99%, FCC, FG
C6H13NO2
I5281
L-Isoleucine, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C6H13NO2
I2752
L-Isoleucine, reagent grade, ≥98% (HPLC)
C6H13NO2
PHR1099
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H13NO2
I7403
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H13NO2
58879
L-Isoleucine, BioUltra, ≥99.5% (NT)
C6H13NO2
56241
L-Isoleucine, certified reference material, TraceCERT®
C6H13NO2