EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Effervescent-salt-assisted dispersive micro-solid-phase extraction using mesoporous hybrid materials coupled with ultra-performance liquid chromatography for the determination of trace-level compounds in complicated plant preparations.
PMID 25527130
Abstract
A novel effervescent-salt-assisted dispersive micro-solid-phase extraction using mesoporous hybrid materials was developed for the extraction of minute traces of constituents in complicated plant preparations. In this study, a special tablet containing carbon dioxide sources (sodium dihydrogenphosphate and sodium carbonate) and the sorbent (mesoporous hybrid materials) was prepared. The effects of different parameters influencing the extraction efficiency such as the concentration of salts, the type and concentration of mesoporous material, pH of diluent, and desorption solvents were investigated and optimized. Results show that the proposed method using green solvents (water) as extraction solutions required low consumption of sample amount and obtained high enrichment efficiency. Moreover, under optimized conditions, the tested tanshinones exhibited good linearity (r(2) > 0.994) in the concentration range of 0.5 to 80 ng mL(-1). The limits of detection values were lower than 0.0803 pg using UV-visible detection. The developed method was successfully applied for the analysis of trace tanshinones in compound Danshen dripping pill and Danqi tablet samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL, analytical standard
C19H30O2
71251
Acetic acid, analytical standard
C2H4O2
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
09684
Ammonia, puriss., anhydrous, ≥99.9%
H3N
294993
Ammonia, anhydrous, ≥99.98%
H3N
779423
Ammonia solution, 4 M in methanol
H3N
718939
Ammonia solution, 0.4 M in THF
H3N
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O
1424109
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
CH4O
03366
Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-, 1.00 g/L, for 1L standard solution, analytical standard
H3O4P
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N