EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
Palmitoleic acid is elevated in fatty liver disease and reflects hepatic lipogenesis.
PMID 25527748
Abstract
Biochemical evidence has linked the coordinate control of fatty acid (FA) synthesis with the activity of stearoyl-CoA desaturase-1 (SCD1). The ratio of 16:1n-7 to 16:0 [SCD1₁₆] in plasma triacylglycerol FA has been used as an index to reflect liver SCD1₁₆ activity and has been proposed as a biomarker of FA synthesis, although this use has not been validated by comparison with isotopically measured de novo lipogenesis (DNL(Meas)). We investigated plasma lipid 16:1n-7 and FA indexes of elongation and desaturation in relation to lipogenesis. In this cross-sectional investigation of metabolism, 24 overweight adults, who were likely to have elevated DNL, consumed D2O for 10 d and had liver fat (LF) measured by magnetic resonance spectroscopy. Very-low-density lipoprotein (VLDL)-triacylglycerols and plasma free FA [nonesterified fatty acids (NEFAs)] were analyzed by using gas chromatography for the FA composition (molar percentage) and gas chromatography-mass spectrometry and gas chromatography-combustion isotope ratio mass spectrometry for deuterium enrichment. In all subjects, VLDL-triacylglycerol 16:1n-7 was significantly (P < 0.01) related to DNL(Meas) (r = 0.56), liver fat (r = 0.53), and adipose insulin resistance (r = 0.56); similar positive relations were shown with the SCD1₁₆ index, and the pattern in NEFAs echoed that of VLDL-triacylglycerols. Compared with subjects with low LF (3.1 ± 2.7%; n = 11), subjects with high LF (18.4 ± 3.6%; n = 13) exhibited a 45% higher VLDL-triacylglycerol 16:1n-7 molar percentage (P < 0.01), 16% of subjects had lower 18:2n-6 (P = 0.01), and 27% of subjects had higher DNL as assessed by using a published DNL index (ratio of 16:0 to 18:2n-6; P = 0.03), which was isotopically confirmed by DNL(Meas) (increased 2.5-fold; P < 0.01). Compared with 16:0 in the diet, the low amount of dietary 16:1n-7 in VLDL-triacylglycerols corresponded to a stronger signal of elevated DNL. The current data provide support for the use of the VLDL-triacylglycerol 16:1n-7 molar percentage as a biomarker for elevated liver fat when isotope use is not feasible; however, larger-scale confirmatory studies are needed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9417
Palmitoleic acid, ≥98.5% (GC), liquid
C16H30O2
76169
Palmitoleic acid, analytical standard
C16H30O2