EMAIL THIS PAGE TO A FRIEND
Breast Cancer Research and Treatment
A preclinical evaluation of the PI3K alpha/delta dominant inhibitor BAY 80-6946 in HER2-positive breast cancer models with acquired resistance to the HER2-targeted therapies trastuzumab and lapatinib.
PMID 25528022
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5011
5′-Deoxy-5′-(methylthio)adenosine
C11H15N5O3S