EMAIL THIS PAGE TO A FRIEND
BMC microbiology
Identification of an immuno-dominant protein from Klebsiella pneumoniae strains causing pyogenic liver abscess: implication in serodiagnosis.
PMID 25528354
Abstract
Klebsiella pneumoniae has emerged worldwide as a cause of pyogenic liver abscess (PLA) often complicated by meningitis and endophthalmitis. Early detection of this infectious disease will improve its clinical outcome. Therefore, we tried to isolate immunodominant proteins secreted by K. pneumoniae strains causing PLA. The secreted proteins of the NTUH-K2044 strain were separated by two-dimensional electrophoresis and then immunoblotted using convalescent sera from patients with K. pneumoniae PLA. A ~30-kDa immunodominant protein was then identified. Liquid chromatography-tandem mass spectrometry (LC-MS/MS) revealed an open reading frame (KP1_p307) located on the pK2044 plasmid and bioinformatic analysis identified this protein as a signal peptide of unknown function. The KP1_p307 gene was more prevalent in PLA strains and capsular type K1/K2 strains, but disruption of this gene in NTUH-K2044 strain did not decrease virulence in mice. Ten of fourteen (71%) sera from patients with K. pneumoniae PLA were immunoreactive with the recombinant KP1_p307 protein. Seroconversion demonstrated by a rise in serum titer in serial serum samples confirmed that antibodies against the KP1_p307 protein were elicited after infection. The KP1_p307 protein could be used as an antigen for early serodiagnosis of K. pneumoniae PLA, particularly in K1/K2 PLA strains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235202
4-Bromomethyl-7-methoxycoumarin, 97%
C11H9BrO3