EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and Environmental Safety
Study on the correlation between copper oxide nanoparticles induced growth suppression and enhanced lignification in Indian mustard (Brassica juncea L.).
PMID 25528486