EMAIL THIS PAGE TO A FRIEND
European Journal of Immunology
TLR9 and STING agonists synergistically induce innate and adaptive type-II IFN.
PMID 25529558
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5817
DMXAA, ≥98% (HPLC), solid
C17H14O4