EMAIL THIS PAGE TO A FRIEND
Food chemistry
Residues investigation of fluoroquinolones and sulphonamides and their metabolites in bovine milk by quantification and confirmation using ultra-performance liquid chromatography-tandem mass spectrometry.
PMID 25529725
Abstract
A novel UPLC-MS/MS method for quantification and confirmation of eight fluoroquinolones, five sulphonamides (SAs) and four acetyled metabolites in milk is developed. Their main fragmentation pathways were presented. The limits of quantification were in the range of 0.01-0.29 μg kg(-1) for FQs and 0.13-1.68 μg kg(-1) for SAs. Mean recoveries ranging from 61% to 115% were achieved at spiked level of 0.5-100 μg kg(-1). The intra- and inter-day precisions were in the range of 6.3-10.7% and 9.0-13.4%, respectively. This novel approach has high speed and sensitivity, and can ensure the multiclass and multiresidue analysis of bovine milk at low μg kg(-1) level. The content of each analyte residue is much lower than the maximum residue limits. Some residues in milk were confirmed in according to the Commission Decision 657/2002/EC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17849
Enrofloxacin, ≥98.0% (HPLC)
C19H22FN3O3
33699
Enrofloxacin, VETRANAL, analytical standard
C19H22FN3O3
Y0001259
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
1235900
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
C19H22FN3O3
PHR1513
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H22FN3O3
Y0001160
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
N9890
Norfloxacin, analytical standard, ≥98% (TLC)
C16H18FN3O3
33899
Norfloxacin, VETRANAL, analytical standard
C16H18FN3O3
N1230000
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001301
Norfloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001015
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
O8757 Ofloxacin, fluoroquinolone antibiotic
C18H20FN3O4
33703
Ofloxacin, VETRANAL, analytical standard
C18H20FN3O4
1478108
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
C18H20FN3O4
PHR1168 Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C18H20FN3O4
S8626
Sulfadiazine, ≥99.0%
C10H10N4O2S
35033
Sulfadiazine, VETRANAL, analytical standard
C10H10N4O2S
S1800000
Sulfadiazine, European Pharmacopoeia (EP) Reference Standard
C10H10N4O2S
1625009
Sulfadiazine, United States Pharmacopeia (USP) Reference Standard
C10H10N4O2S
PHR1343
Sulfadiazine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H10N4O2S
Y0001480
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
C10H10N4O2S
S7007
Sulfadimethoxine, analytical standard, ≥98.5% (TLC)
C12H14N4O4S
46794
Sulfadimethoxine, VETRANAL, analytical standard
C12H14N4O4S
PHR1448
Sulfadimethoxine
C12H14N4O4S
Y0001679
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
C12H14N4O4S
S1900000
Sulfadimidine, European Pharmacopoeia (EP) Reference Standard
C12H14N4O2S
Y0001495
Sulfadimidine for peak identification, European Pharmacopoeia (EP) Reference Standard
C12H14N4O2S
S8876
Sulfamerazine, ReagentPlus®, ≥99.0%
C11H12N4O2S
46826
Sulfamerazine, VETRANAL, analytical standard
C11H12N4O2S
S2000000
Sulfamerazine, European Pharmacopoeia (EP) Reference Standard
C11H12N4O2S