EMAIL THIS PAGE TO A FRIEND
Placenta
Pharmacological inhibition of MIF interferes with trophoblast cell migration and invasiveness.
PMID 25530499
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43720
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
C9H8N2O7