EMAIL THIS PAGE TO A FRIEND
Singapore medical journal
Production of a fragment of glycoprotein G of herpes simplex virus type 2 and evaluation of its diagnostic potential.
PMID 25532518
Abstract
Herpes simplex virus type 2 (HSV-2) is the most common cause of genital herpes. Glycoprotein G (gG) is a prototype antigen for type-specific serodiagnosis distinguishing between HSV type 1 (HSV-1) and HSV-2 infections. As immunological diagnosis kits for accurate differentiation between HSV-1 and HSV-2 antibodies can be expensive, there is a need to develop a convenient, sensitive, specific and cost-effective serodiagnostic kit. We successfully expressed a fragment of gG comprising residues 321-580 of HSV-2 with histidine tag (gG(321-580His)) in a Bac-to-Bac baculovirus expression system, which had an antigenicity similar to its native counterpart. An indirect enzyme-linked immunosorbent assay (ELISA) was developed using gG(321-580His) as the diagnostic antigen and evaluated by comparison with a commercial HerpeSelect 2 ELISA immunoglobulin G kit as reference. In testing 318 field serum samples, the diagnostic relative sensitivity and specificity of the developed gG(321-580His)-ELISA test in qualitative comparison with the commercial kit were 93.81% and 96.74%, respectively, and the accuracy was 94.65%. The study indicates that gG(321-580His) has a high diagnostic potential for HSV-2 virus serodiagnosis in humans.