EMAIL THIS PAGE TO A FRIEND
Molecular Genetics and Metabolism
Intravenous injection of AAVrh10-GALC after the neonatal period in twitcher mice results in significant expression in the central and peripheral nervous systems and improvement of clinical features.
PMID 25533112