EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research. Part B, Applied Biomaterials
Vertical bone augmentation induced by ultrathin hydroxyapatite sputtered coated mini titanium implants in a rabbit calvaria model.
PMID 25533173