EMAIL THIS PAGE TO A FRIEND
Vaccine
Protection of non-human primates against glanders with a gold nanoparticle glycoconjugate vaccine.
PMID 25533326
Abstract
The Gram-negative Burkholderia mallei is a zoonotic pathogen and the causative agent of glanders disease. Because the bacteria maintain the potential to be used as a biothreat agent, vaccine strategies are required for human glanders prophylaxis. A rhesus macaque (Macaca mulatta) model of pneumonic (inhalational) glanders was established and the protective properties of a nanoparticle glycoconjugate vaccine composed of Burkholderia thailandensis LPS conjugated to FliC was evaluated. An aerosol challenge dose of ∼1×10(4) CFU B. mallei produced mortality in 50% of naïve animals (n=2/4), 2-3 days post-exposure. Although survival benefit was not observed by vaccination with a glycoconjugate glanders vaccine (p=0.42), serum LPS-specific IgG titers were significantly higher on day 80 in 3 vaccinated animals who survived compared with 3 vaccinated animals who died. Furthermore, B. mallei was isolated from multiple organs of both non-vaccinated survivors, but not from any organs of 3 vaccinated survivors at 30 days post-challenge. Taken together, this is the first time a candidate vaccine has been evaluated in a non-human primate aerosol model of glanders and represents the initial step for consideration in pre-clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9043
S-Acetylthioglycolic acid N-hydroxysuccinimide ester, ≥95% (TLC), powder
C8H9NO5S