EMAIL THIS PAGE TO A FRIEND
Toxins
Protective effect of two yeast based feed additives on pigs chronically exposed to deoxynivalenol and zearalenone.
PMID 25533517
Abstract
To evaluate the effects of the mycotoxins deoxynivalenol (DON) and zearalenone (ZEA) on pigs and the benefits of two mycotoxin mitigation strategies, gilts (n = 84, 9.1 ± 0.1 kg) were allotted to four treatments: CON (control); MT (4.8 mg/kg feed DON and 0.3 mg/kg feed ZEA); MT-YC (MT + 2 g/kg of yeast cell wall product); and MT-YF (MT + 2 g/kg of yeast fermentation product). After 42 days of feeding, pigs fed MT had reduced (p < 0.05) growth performance compared with pigs fed CON. Pigs fed MT-YF had greater (p < 0.05) average daily gain and tended to have greater (p = 0.080) average daily feed intake than MT, whereas pigs fed MT-YC did not differ from MT. Oxidative DNA damage increased (p < 0.05) in MT, whereas pigs fed MT-YF tended to have lower (p = 0.067) oxidative stress. Liver hydropic degeneration was increased (p < 0.05) in MT in contrast to CON and MT-YF, and tended to be greater (p = 0.079) than MT-YC. Collectively, feeding diets contaminated with mycotoxins significantly reduced growth performance and impacted pig health. The yeast additives had varied ability to reduce mycotoxin effects on pig growth and health, but may still play a beneficial role in reducing the overall impacts of a mycotoxin challenge on pigs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6636
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
C17H12O6
03372
Copper atomic spectroscopy standard concentrate 10.00 g Cu, 10.00 g/L, for 1L standard solution, analytical standard
Cl2Cu
C3000000
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
C63H88CoN14O14P
PHR1234
Cyanocobalamin, Pharmaceutical Secondary Standard; Certified Reference Material
C63H88CoN14O14P
BP466
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
C63H88CoN14O14P
32943
Deoxynivalenol, reference material
C15H20O6
1381006
Menadione, United States Pharmacopeia (USP) Reference Standard
C11H8O2
M0300000
Menadione, European Pharmacopoeia (EP) Reference Standard
C11H8O2
PHR1411
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
C11H8O2
R0300000
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
C22H32O2
1716002
Retinyl acetate, United States Pharmacopeia (USP) Reference Standard
C22H32O2
R4632
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
C22H32O2
R7882
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
C22H32O2
R3250
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
C22H32O2
95140
Retinyl acetate, concentrate (~50% (w/w) in peanut oil), ~1500 U/mg
C22H32O2
46958
Retinyl acetate, analytical standard
C22H32O2
PHR1236
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
C22H32O2
PHR1054
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N4O6
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
R0600000
Riboflavin, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
Y0000757
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
V7763
Vitamin A (acetate), meets USP testing specifications
C22H32O2