EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Patterns of genome evolution that have accompanied host adaptation in Salmonella.
PMID 25535353
Abstract
Many bacterial pathogens are specialized, infecting one or few hosts, and this is often associated with more acute disease presentation. Specific genomes show markers of this specialization, which often reflect a balance between gene acquisition and functional gene loss. Within Salmonella enterica subspecies enterica, a single lineage exists that includes human and animal pathogens adapted to cause infection in different hosts, including S. enterica serovar Enteritidis (multiple hosts), S. Gallinarum (birds), and S. Dublin (cattle). This provides an excellent evolutionary context in which differences between these pathogen genomes can be related to host range. Genome sequences were obtained from ∼ 60 isolates selected to represent the known diversity of this lineage. Examination and comparison of the clades within the phylogeny of this lineage revealed signs of host restriction as well as evolutionary events that mark a path to host generalism. We have identified the nature and order of events for both evolutionary trajectories. The impact of functional gene loss was predicted based upon position within metabolic pathways and confirmed with phenotyping assays. The structure of S. Enteritidis is more complex than previously known, as a second clade of S. Enteritidis was revealed that is distinct from those commonly seen to cause disease in humans or animals, and that is more closely related to S. Gallinarum. Isolates from this second clade were tested in a chick model of infection and exhibited a reduced colonization phenotype, which we postulate represents an intermediate stage in pathogen-host adaptation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
134368
1,2-Propanediol, ReagentPlus®, 99%
C3H8O2
398039
1,2-Propanediol, ACS reagent, ≥99.5%
C3H8O2
16033
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
C3H8O2
82280
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C3H8O2
12279
1,2-Propanediol, analytical standard
C3H8O2
82281
1,2-Propanediol, tested according to Ph Eur
C3H8O2
55561
Aucubin, analytical standard
C15H22O9
00090590
Aucubin, primary reference standard
C15H22O9
411000
Ethanolamine, purified by redistillation, ≥99.5%
C2H7NO
E9508
Ethanolamine, ≥98%
C2H7NO
398136
Ethanolamine, ACS reagent, ≥99.0%
C2H7NO
15014
Ethanolamine, ≥99%
C2H7NO
ARK2169
Ethanolamine, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for Pharma or biopharmaceutical production
C2H7NO
E0135
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
C2H7NO
02400
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
C2H7NO
00278
Ethanolamine, analytical standard
C2H7NO
110167
Ethanolamine, ReagentPlus®, ≥99%
C2H7NO
W294004
Propylene glycol, FCC, FG
C3H8O2
PHR1051
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O2
1576708
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
C3H8O2
Y0001547
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
C3H8O2
P4347
Propylene Glycol, meets USP testing specifications
C3H8O2
Y0001184
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
C2H7NO