EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Hooking She3p onto She2p for myosin-mediated cytoplasmic mRNA transport.
PMID 25535369
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34138
Neosolaniol solution, 100 μg/mL in acetonitrile, analytical standard
C19H26O8