EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
B-cell repertoire responses to varicella-zoster vaccination in human identical twins.
PMID 25535378
Abstract
Adaptive immune responses in humans rely on somatic genetic rearrangements of Ig and T-cell receptor loci to generate diverse antigen receptors. It is unclear to what extent an individual's genetic background affects the characteristics of the antibody repertoire used in responding to vaccination or infection. We studied the B-cell repertoires and clonal expansions in response to attenuated varicella-zoster vaccination in four pairs of adult identical twins and found that the global antibody repertoires of twin pair members showed high similarity in antibody heavy chain V, D, and J gene segment use, and in the length and features of the complementarity-determining region 3, a major determinant of antigen binding. These twin similarities were most pronounced in the IgM-expressing B-cell pools, but were seen to a lesser extent in IgG-expressing B cells. In addition, the degree of antibody somatic mutation accumulated in the B-cell repertoire was highly correlated within twin pair members. Twin pair members had greater numbers of shared convergent antibody sequences, including mutated sequences, suggesting similarity among memory B-cell clonal lineages. Despite these similarities in the memory repertoire, the B-cell clones used in acute responses to ZOSTAVAX vaccination were largely unique to each individual. Taken together, these results suggest that the overall B-cell repertoire is significantly shaped by the underlying germ-line genome, but that stochastic or individual-specific effects dominate the selection of clones in response to an acute antigenic stimulus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09743
3,3′,5,5′-Tetramethylbenzidine, standard for GC
C16H20N2