EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Transcriptomic identification and expression of starch and sucrose metabolism genes in the seeds of Chinese chestnut (Castanea mollissima).
PMID 25537355
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52445
Anthrone, analytical standard
C14H10O
319899
Anthrone, ACS reagent, 97%
C14H10O