EMAIL THIS PAGE TO A FRIEND
Natural product research
Ascomycotin A, a new citromycetin analogue produced by Ascomycota sp. Ind19F07 isolated from deep sea sediment.
PMID 25537370
Abstract
A new citromycetin analogue, ascomycotin A (1), together with eight known compounds, wortmannilactone E (2), orcinol (3), orsellinic acid (4), isosclerone (5), (3R,4S)-( - )-4-hydroxymellein (6), diorcinol (7), chaetocyclinone B (8) and 2,5-dimethoxy-3,6-di(p-methoxypheny1)-1,4-benzoquinone (9), was isolated from the fungal strain Ascomycota sp. Ind19F07, which was isolated from the deep sea sediment of the Indian Ocean. The structures of the compounds were established by spectroscopic data including 1D and 2D NMR and HR-ESI-MS. Compounds (1-9) were evaluated for antibacterial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447420
Orcinol, 97%
C7H8O2