EMAIL THIS PAGE TO A FRIEND
Nuclear Medicine and Biology
Development of 4-hexadecyl-4,7-diaza-1,10-decanedithiol (HDD) kit for the preparation of the liver cancer therapeutic agent Re-188-HDD/lipiodol.
PMID 25537725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238279
1-Iodohexadecane, contains copper as stabilizer, 95%
C16H33I