EMAIL THIS PAGE TO A FRIEND
Nuclear Medicine and Biology
In vivo imaging and characterization of [(18)F]DPA-714, a potential new TSPO ligand, in mouse brain and peripheral tissues using small-animal PET.
PMID 25537727