EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Renal Physiology
Sex differences in renal and metabolic responses to a high-fructose diet in mice.
PMID 25537743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1462
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
C4H7N3O
F0127
D-(−)-Fructose, ≥99%
C6H12O6
F2543
D-(−)-Fructose, BioXtra
C6H12O6
F3510
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
F9048
D-(−)-Fructose, meets USP testing specifications
C6H12O6
F2793
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
C6H12O6
47739
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
C6H12O6
47740
D-(−)-Fructose, for microbiology, ≥99.0%
C6H12O6
47748
D-(−)-Fructose, tested according to Ph.Eur.
C6H12O6
12156
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
H2O
1286504
Fructose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
F0550000
Fructose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
PHR1002
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
DENWAT
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
DENWAT3
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
U2625
Uric acid, ≥99%, crystalline
C5H4N4O3
U0881
Uric acid, BioXtra, ≥99% (HPLC)
C5H4N4O3
14211
Water, ACS reagent, for ultratrace analysis
H2O
95283
Water, for HPCE, for luminescence, for UV-spectroscopy
H2O
99053
Water, deionized
H2O
95280
Water, tested according to Ph.Eur.
H2O
78533
Water, for TOC analysis
H2O
00612
Water, for ion chromatography
H2O
38155
Water with 0.1% trifluoroacetic acid
H2O