EMAIL THIS PAGE TO A FRIEND
AJR. American journal of roentgenology
Colonic distention at CT colonography: randomized evaluation of both IV hyoscine butylbromide and automated carbon dioxide insufflation.
PMID 25539240
Abstract
The purpose of this article is to evaluate the efficacy of IV hyoscine butylbromide as a bowel relaxant and automated carbon dioxide insufflation in CT colonography in terms of colonic distention and perceived burden. SUBJECTS AND METHODS; Two hundred twenty-four participants were randomly allocated to one of four groups: control (no bowel relaxant and IV saline placebo before CT colonography with manual carbon dioxide insufflation), hyoscine butylbromide (IV hyoscine butylbromide before examination with manual carbon dioxide insufflation), automated (no bowel relaxant before examination with automated carbon dioxide insufflation), and combined (hyoscine butylbromide before examination with automated carbon dioxide insufflation). The degree of colonic distention on a 4-point scale, examination time, and participants' satisfaction, as measured by their responses to a questionnaire, were assessed. The mean distention grades of all the colonic segments and both positions were 3.22 in the control group, 3.28 in the hyoscine butylbromide group, 3.77 in the automated group, and 3.74 in the combined group. Compared with manual carbon dioxide insufflation, automated carbon dioxide insufflation significantly improved the clinical adequacy of colonic distention and shortened examination time. No statistically significant difference was seen in the clinical adequacy of distention between participants who received hyoscine butylbromide and those who did not, or in examination time. Overall, the participants' experiences were not different. Colonic distention was statistically significantly improved by automated carbon dioxide insufflation, but not by the administration of hyoscine butylbromide. The participants' tolerance was similar in each group.