EMAIL THIS PAGE TO A FRIEND
International journal of radiation oncology, biology, physics
Preclinical evaluation of bioabsorbable polyglycolic acid spacer for particle therapy.
PMID 25539373
Abstract
To evaluate the efficacy and safety of a polyglycolic acid (PGA) spacer through physical and animal experiments. The spacer was produced with surgical suture material made of PGA, forming a 3-dimensional nonwoven fabric. For evaluation or physical experiments, 150-MeV proton or 320-MeV carbon-ion beams were used to generate 60-mm width of spread-out Bragg peak. For animal experiments, the abdomens of C57BL/6 mice, with or without the inserted PGA spacers, were irradiated with 20 Gy of carbon-ion beam (290 MeV) using the spread-out Bragg peak. Body weight changes over time were scored, and radiation damage to the intestine was investigated using hematoxylin and eosin stain. Blood samples were also evaluated 24 days after the irradiation. Long-term thickness retention and safety were evaluated using crab-eating macaques. No chemical or structural changes after 100 Gy of proton or carbon-ion irradiation were observed in the PGA spacer. Water equivalency of the PGA spacer was equal to the water thickness under wet condition. During 24 days' observation after 20 Gy of carbon-ion irradiation, the body weights of mice with the PGA spacer were relatively unchanged, whereas significant weight loss was observed in those mice without the PGA spacer (P<.05). In mice with the PGA spacer, villus and crypt structure were preserved after irradiation. No inflammatory reactions or liver or renal dysfunctions due to placement of the PGA spacer were observed. In the abdomen of crab-eating macaques, thickness of the PGA spacer was maintained 8 weeks after placement. The absorbable PGA spacer had water-equivalent, bio-compatible, and thickness-retaining properties. Although further evaluation is warranted in a clinical setting, the PGA spacer may be effective to stop proton or carbon-ion beams and to separate normal tissues from the radiation field.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
05105
Activated charcoal, powder
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
31616
Activated charcoal, puriss. p.a., powder
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
675326
Activated charcoal, DARCO® KB-G
C
05112
Activated charcoal, for GC
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
C7606
Activated Charcoal, meets USP testing specifications
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
97876
Activated Charcoal Norit®, Norit CA1, wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF23879009
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
C
GF34902109
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
C
GF24640950
Carbon, rod, 100mm, diameter 10.0mm, graphite, 100%
C
GF01171763
Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%
C