EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Sequence-dependent off-target inhibition of TLR7/8 sensing by synthetic microRNA inhibitors.
PMID 25539920
Abstract
Anti-microRNA (miRNA) oligonucleotides (AMOs) with 2'-O-Methyl (2'OMe) residues are commonly used to study miRNA function and can achieve high potency, with low cytotoxicity. Not withstanding this, we demonstrate the sequence-dependent capacity of 2'OMe AMOs to inhibit Toll-like receptor (TLR) 7 and 8 sensing of immunostimulatory RNA, independent of their miRNA-targeting function. Through a screen of 29 AMOs targeting common miRNAs, we found a subset of sequences highly inhibitory to TLR7 sensing in mouse macrophages. Interspecies conservation of this inhibitory activity was confirmed on TLR7/8 activity in human peripheral blood mononuclear cells. Significantly, we identified a core motif governing the inhibitory activity of these AMOs, which is present in more than 50 AMOs targeted to human miRNAs in miRBaseV20. DNA/locked nucleic acids (LNA) AMOs synthesized with a phosphorothioate backbone also inhibited TLR7 sensing in a sequence-dependent manner, demonstrating that the off-target effects of AMOs are not restricted to 2'OMe modification. Taken together, our work establishes the potential for off-target effects of AMOs on TLR7/8 function, which should be taken into account in their therapeutic development and in vivo application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12156
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
H2O
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2
P1190
Potassium acetate, for molecular biology, ≥99.0%
C2H3KO2
P5708
Potassium acetate, BioXtra, ≥99.0%
C2H3KO2
P1222
Potassium acetate, meets USP testing specifications
C2H3KO2
255785
Potassium acetate, 99.98% trace metals basis, ~100 ppm Rb, <50 ppm Cs
C2H3KO2
236497
Potassium acetate, ACS reagent, ≥99.0%
C2H3KO2
P1147
Potassium acetate, ReagentPlus®, ≥99.0%
C2H3KO2
60035
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
C2H3KO2
32309
Potassium acetate, puriss. p.a., ≥99% (perchloric acid titration)
C2H3KO2
25059
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
C2H3KO2
W292001
Potassium acetate, 98%
C2H3KO2
95843
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
C2H3KO2
DENWAT
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
DENWAT3
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
14211
Water, ACS reagent, for ultratrace analysis
H2O
95283
Water, for HPCE, for luminescence, for UV-spectroscopy
H2O
99053
Water, deionized
H2O
95280
Water, tested according to Ph Eur
H2O
78533
Water, for TOC analysis
H2O
00612
Water, for ion chromatography
H2O
38155
Water with 0.1% trifluoroacetic acid
H2O