EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Analysis of psilocin, bufotenine and LSD in hair.
PMID 25540060
Abstract
A method for the simultaneous extraction of the hallucinogens psilocin, bufotenine, lysergic acid diethylamide (LSD) as well as iso-LSD, nor-LSD and O-H-LSD from hair with hydrochloride acid and methanol is presented. Clean-up of the hair extracts is performed with solid phase extraction using a mixed-mode cation exchanger. Extracts are measured with liquid chromatography coupled with electrospray tandem mass spectrometry. The method was successfully validated according to the guidelines of the 'Society of Toxicological and Forensic Chemistry' (GTFCh). To obtain reference material hair was soaked in a solution of the analytes in dimethyl sulfoxide/methanol to allow incorporation into the hair. These fortified hair samples were used for method development and can be employed as quality controls.