EMAIL THIS PAGE TO A FRIEND
Clinica Chimica Acta; International Journal of Clinical Chemistry
Rapid measurement of tacrolimus in whole blood by paper spray-tandem mass spectrometry (PS-MS/MS).
PMID 25540885
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F4679
FK-506 monohydrate, ≥98% (HPLC)
C44H69NO12 · H2O
1642802
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
C44H69NO12 · H2O