PLoS ONE
The roles of co-chaperone CCRP/DNAJC7 in Cyp2b10 gene activation and steatosis development in mouse livers.
PMID 25542016
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9568
3,3′-Diindolylmethane, ≥98% (HPLC)
C17H14N2
D6513
Desmosterol, ≥84% (GC)
C27H44O