EMAIL THIS PAGE TO A FRIEND
The International Journal of Biochemistry & Cell Biology
The linker histone in Saccharomyces cerevisiae interacts with actin-related protein 4 and both regulate chromatin structure and cellular morphology.
PMID 25542182
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1423
Sodium Nitroprusside, Pharmaceutical Secondary Standard; Certified Reference Material
C5FeN6Na2O · 2H2O