EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Fast agitated directly suspended droplet microextraction technique for the rapid analysis of eighteen organophosphorus pesticides in human blood.
PMID 25542703
Abstract
A new sample preparation technique named as fast agitated directly suspended droplet microextraction (FA-DSDME) was proposed as an improved version of directly suspended droplet microextraction (DSDME) for the extraction and pre-concentration of wide-range organophosphorus pesticides (OPPs) from human blood prior to liquid chromatography tandem mass spectrometric (LC-MS/MS) analysis. In this method, instead of protecting the unwanted rupturing of extraction droplet (organic solvent), it was deliberately splintered into fine droplets by providing automated high-speed agitation to the biphasic extraction system (extraction solvent and sample solution). Fine organic droplets were then recollected into one, not by using a centrifuge machine but just by giving a very slow stirring to the bottom of the extraction system. The present method has surmounted the problem of prolonged extraction time associated with old DSDME. Under optimum extraction conditions, the method showed good sensitivity with low detection limits ranging from 0.0009 to 0.122μgL(-1). Mean recoveries were achieved in the range of 86-109% at three levels of spiking concentration (low, middle and high) from linearity range of individual analyte. Intra-day and inter-day precisions were ≤4.68 and ≤9.57 (%RSD) respectively. Enrichment factor (EF) for each analyte varied from 30 to 132 which prove the ability of this technique to pre-concentrate the extracted analytes up to a good extent. The sample matrices have shown an insignificant influence on method's sensitivity. The proposed method may find immense use in epidemiological, toxicological, regulatory and forensic laboratories.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44095
1-Dodecanol, Selectophore, ≥98.0%
C12H26O
443816
1-Dodecanol, ACS reagent, ≥98.0%
C12H26O
126799
1-Dodecanol, reagent grade, 98%
C12H26O
75544
1-Dodecanol, analytical standard
C12H26O
U1001
1-Undecanol, 99%
C11H24O
D221503
2-Dodecanol, 99%
C12H26O
45395
Chlorpyrifos, PESTANAL®, analytical standard
C9H11Cl3NO3PS
94114
Chlorpyrifos, certified reference material, TraceCERT®
C9H11Cl3NO3PS
45441
Dichlorvos, PESTANAL®, analytical standard
C4H7Cl2O4P
45449
Dimethoate, PESTANAL®, analytical standard
C5H12NO3PS2
442573
Dursban, analytical standard
C9H11Cl3NO3PS
45477
Ethion, PESTANAL®, analytical standard
C9H22O4P2S4
N11842
Fonofos, analytical standard
C10H15OPS2
33587
Fonofos solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C10H15OPS2
52209
Hexadecane, analytical standard
C16H34
296317
Hexadecane, anhydrous, ≥99%
C16H34
442679
Hexadecane, analytical standard
C16H34
H6703
Hexadecane, ReagentPlus®, 99%
C16H34
W261718
Lauryl alcohol, ≥98%, FG
C12H26O
36143
Malathion, PESTANAL®, analytical standard
C10H19O6PS2
91481
Malathion, certified reference material, TraceCERT®
C10H19O6PS2
1374408
Malathion, United States Pharmacopeia (USP) Reference Standard
C10H19O6PS2
SCM-039
Malathion, vial of 1 g, analytical standard
C10H19O6PS2
M0120000
Malathion, European Pharmacopoeia (EP) Reference Standard
C10H19O6PS2
N12397
Methidathion, analytical standard
C6H11N2O4PS3
36158
Methidathion, PESTANAL®, analytical standard
C6H11N2O4PS3
PS679
Methidathion, analytical standard
C6H11N2O4PS3
36173
Monocrotophos, PESTANAL®, analytical standard
C7H14NO5P
D9286
Paraoxon-ethyl, ≥90%, oil
C10H14NO6P
36186
Paraoxon-ethyl, PESTANAL®, analytical standard
C10H14NO6P