EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The uptake of HIV Tat peptide proceeds via two pathways which differ from macropinocytosis.
PMID 25542781
Abstract
Cell penetrating peptides (CPPs) have been extensively studied as vectors for cellular delivery of therapeutic molecules, yet the identity of their uptake routes remained unclear and is still under debate. In this study we provide new insights into CPP entry routes by quantitatively measuring the intracellular uptake of FAM-labeled Tat-peptide under rigorous kinetic and thermal conditions. The uptake of Tat-peptide between 4 and 15°C corresponds to Q10=1.1, proceeding through a prompt (<5 min), temperature-independent process, suggesting direct membrane translocation. At longer durations, Tat rate of uptake shows linear dependence on temperature with Q10=1.44, accompanied by activation energy Ea=4.45 Kcal/mole. These values are significantly lower than those we found for the macropinocytosis probe dextran (Q10=2.2 and Ea=7.2 Kcal/mole) which possesses an exponential dependence on temperature, characteristic of endocytosis processes. Tat-peptide and dextran do not interfere with each other's uptake rate and the ratio of Tat-peptide uptake to its extracellular concentration is ~15 times higher than that for dextran. In addition, Phloretin, a modulator of cell membrane dipole potential, is shown to increase dextran uptake but to reduce that of Tat. We conclude that the uptake of Tat differs from that of dextran in all parameters. Tat uptake proceeds by dual entry routes which differ by their energy dependence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1250
6-Ketocholestanol
C27H46O2
P7912
Phloretin, ≥99%
C15H14O5
91897
Phloretin, analytical standard
C15H14O5