EMAIL THIS PAGE TO A FRIEND
Journal of hepatology
Pharmacological activation of aldehyde dehydrogenase 2 by Alda-1 reverses alcohol-induced hepatic steatosis and cell death in mice.
PMID 25543082
Abstract
Effective therapies for alcoholic liver disease are currently unavailable. The present study tested the efficacy of Alda-1, a specific aldehyde dehydrogenase 2 (ALDH2) activator, in treating alcoholic liver disease. Male C57BL/6J mice were exposed to alcohol for a time-course study on aldehyde metabolism. The specificity and efficacy of Alda-1 on activating hepatic ALDH2 and aldehyde clearance were determined by acute treatments. Then, mice were fed alcohol for 8 weeks with Alda-1 administration for the last 10 days to test the therapeutic potential of Alda-1. Lastly, H4IIEC3 cells were treated with ethanol, acetaldehyde, or 4-hydroxynonenal to define the link between aldehydes and hepatotoxicity. Alcohol feeding for 8 weeks induced hepatic ALDH2 dysfunction and aldehyde accumulation. One dose of Alda-1 administration elevated hepatic ALDH activity, which was blocked by the specific ALDH2 inhibitor, daidzin. Alda-1 accelerated acetaldehyde clearance after acute alcohol intoxication. Alda-1 treatment in the 8-week alcohol feeding model reversed liver damage along with reduction of hepatic aldehydes. Alda-1 re-activated transcription factors, upregulated fatty acid oxidation enzymes, and reversed steatosis. Alcohol-induced endoplasmic reticulum stress and apoptotic cell death were also attenuated by Alda-1. Acetaldehyde or 4-hydroxynonenal treatment to H4IIEC3 cells inactivated transcription factors and induced endoplasmic reticulum stress and apoptosis, while ethanol per se showed limited effects. Pharmacological activation of ALDH2 by Alda-1 reversed alcoholic steatosis and apoptosis through accelerating aldehyde clearance. This study indicates that ALDH2 is a promising molecular target and Alda-1 has therapeutic potential for treating alcoholic liver disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0462
Alda-1, ≥98% (HPLC)
C15H11Cl2NO3
30408
Daidzin, ≥95.0% (HPLC)
C21H20O9