EMAIL THIS PAGE TO A FRIEND
Archives of Oral Biology
In vitro effect of Paullinia cupana (guarana) on hydrophobicity, biofilm formation, and adhesion of Candida albicans' to polystyrene, composites, and buccal epithelial cells.
PMID 25543106