EMAIL THIS PAGE TO A FRIEND
Aquatic Toxicology (Amsterdam, Netherlands)
A primary fish gill cell culture model to assess pharmaceutical uptake and efflux: evidence for passive and facilitated transport.
PMID 25544062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0884
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
C16H21NO2 · HCl
40543
(±)-Propranolol hydrochloride, analytical reference material
C16H21NO2 · HCl
V4629
(±)-Verapamil hydrochloride, ≥99% (titration), powder
C27H38N2O4 · HCl
PHR1131
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C27H38N2O4 · HCl
138576
1-Adamantylamine, 97%
C10H17N
C4522
Cimetidine
C10H16N6S
PHR1075
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H16N6S
C2175000
Cimetidine, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
1134062
Cimetidine, United States Pharmacopeia (USP) Reference Standard
C10H16N6S
Y0001130
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
Y0001146
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
C10H16N6S
1576005
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
C16H21NO2 · HCl
P3500000
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
C16H21NO2 · HCl
PHR1308
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C16H21NO2 · HCl
P3600000
Propranolol hydrochloride for performance test, European Pharmacopoeia (EP) Reference Standard
C16H21NO2 · HCl
Q3625
Quinidine, anhydrous
C20H24N2O2
22600
Quinidine, crystallized, ≥98.0% (dried material, NT)
C20H24N2O2
1711202
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
C27H38N2O4 · HCl
V0100000
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
C27H38N2O4 · HCl