EMAIL THIS PAGE TO A FRIEND
European Journal of Medicinal Chemistry
Synthesis of functionalized 3-, 5-, 6- and 8-aminoquinolines via intermediate (3-pyrrolin-1-yl)- and (2-oxopyrrolidin-1-yl)quinolines and evaluation of their antiplasmodial and antifungal activity.
PMID 25544689