EMAIL THIS PAGE TO A FRIEND
Monatshefte fur chemie
(m.n)-Homorubins. Syntheses and Structures.
PMID 25544780
Abstract
Five new homorubin analogs of bilirubin with their two dipyrrinone components conjoined to (CH
Related Materials