PloS one
The role of histamine in the retina: studies on the Hdc knockout mouse.
PMID 25545149
Abstract
The role of histamine in the retina is not well understood, despite it regulating a number of functions within the brain, including sleep, feeding, energy balance, and anxiety. In this study we characterized the structure and function of the retina in mice that lacked expression of the rate limiting enzyme in the formation of histamine, histidine decarboxylase (Hdc-/- mouse). Using laser capture microdissection, Hdc mRNA expression was assessed in the inner and outer nuclear layers of adult C57Bl6J wildtype (WT) and Hdc(-/-)-retinae. In adult WT and Hdc(-/-)-mice, retinal fundi were imaged, retinal structure was assessed using immunocytochemistry and function was probed by electroretinography. Blood flow velocity was assessed by quantifying temporal changes in the dynamic fluorescein angiography in arterioles and venules. In WT retinae, Hdc gene expression was detected in the outer nuclear layer, but not the inner nuclear layer, while the lack of Hdc expression was confirmed in the Hdc-/- retina. Preliminary examination of the fundus and retinal structure of the widely used Hdc-/- mouse strain revealed discrete lesions across the retina that corresponded to areas of photoreceptor abnormality reminiscent of the rd8 (Crb1) mutation. This was confirmed after genotyping and the strain designated Hdcrd8/rd8. In order to determine the effect of the lack of Hdc-alone on the retina, Hdc-/- mice free of the Crb1 mutation were bred. Retinal fundi appeared normal in these animals and there was no difference in retinal structure, macrogliosis, nor any change in microglial characteristics in Hdc-/- compared to wildtype retinae. In addition, retinal function and retinal blood flow dynamics showed no alterations in the Hdc-/- retina. Overall, these results suggest that histamine plays little role in modulating retinal structure and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6126
(R)-(−)-Phenylephrine hydrochloride, powder
C9H13NO2 · HCl
P8155
(R)-(−)-Phenylephrine hydrochloride, analytical standard
C9H13NO2 · HCl
F6377
Fluorescein sodium salt, used as fluorescent tracer
C20H10Na2O5
46960
Fluorescein sodium salt, BioReagent, suitable for fluorescence
C20H10Na2O5
30181
Fluorescein sodium salt, analytical standard
C20H10Na2O5
46970
Fluorescein sodium salt, p.a., for source staining
C20H10Na2O5
44208
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C6H14N2O2 · HCl
PHR1101
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N2O2 · HCl
67448
L-Lysine monohydrochloride, certified reference material, TraceCERT®
C6H14N2O2 · HCl
L0900000
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H14N2O2 · HCl
P5704
Monoclonal Anti-Protein Kinase C (PKC) antibody produced in mouse, clone MC5, ascites fluid
1533002
Phenylephrine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C9H13NO2 · HCl
P1250000
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C9H13NO2 · HCl
PHR1017
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C9H13NO2 · HCl
Y0000683
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
C9H13NO2 · HCl
190780
Sodium trichloroacetate, 97%
C2Cl3NaO2