EMAIL THIS PAGE TO A FRIEND
Cancer Cell International
Effects of suberoylanilide hydroxamic acid (SAHA) combined with paclitaxel (PTX) on paclitaxel-resistant ovarian cancer cells and insights into the underlying mechanisms.
PMID 25546354