EMAIL THIS PAGE TO A FRIEND
Clinical toxicology (Philadelphia, Pa.)
High-dose hydroxocobalamin administered after H2S exposure counteracts sulfide-poisoning-induced cardiac depression in sheep.
PMID 25546714
Abstract
Severe H2S poisoning leads to death by rapid respiratory and cardiac arrest, the latter can occur within seconds or minutes in severe forms of intoxication. To determine the time course and the nature of H2S-induced cardiac arrest and the effects of high-dose hydroxocobalamin administered after the end of sulfide exposure. NaHS was infused in 16 sedated mechanically ventilated sheep to reach concentrations of H2S in the blood, which was previously found to lead to cardiac arrest within minutes following the cessation of H2S exposure. High-dose hydroxocobalamin (5 g) or saline solution was administered intravenously, 1 min after the cessation of NaHS infusion. All animals were still alive at the cessation of H2S exposure. Three animals (18%) presented a cardiac arrest within 90 s and were unable to receive any antidote or vehicle. In the animals that survived long enough to receive either hydroxocobalamin or saline, 71% (5/7) died in the control group by cardiac arrest within 10 min. In all instances, cardiac arrest was the result of a pulseless electrical activity (PEA). In the group that received the antidote, intravenous injection of 5 g of hydroxocobalamin provoked an abrupt increase in blood pressure and blood flow; PEA was prevented in all instances. However, we could not find any evidence for a recovery in oxidative metabolism in the group receiving hydroxocobalamin, as blood lactate remained elevated and even continued to rise after 1 h, despite restored hemodynamics. This, along with an unaltered recovery of H2S kinetics, suggests that hydroxocobalamin did not act through a mechanism of H2S trapping. In this sheep model, there was a high risk for cardiac arrest, by PEA, persisting up to 10 min after H2S exposure. Very high dose of hydroxocobalamin (5 g), injected very early after the cessation of H2S exposure, improved cardiac contractility and prevented PEA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1428000 Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
C62H89CoN13O15P