EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Synthesis of uniform cyclodextrin thioethers to transport hydrophobic drugs.
PMID 25550759
Abstract
Methyl and ethyl thioether groups were introduced at all primary positions of α-, β-, and γ-cyclodextrin by nucleophilic displacement reactions starting from the corresponding per-(6-deoxy-6-bromo)cyclodextrins. Further modification of all 2-OH positions by etherification with iodo terminated triethylene glycol monomethyl ether (and tetraethylene glycol monomethyl ether, respectively) furnished water-soluble hosts. Especially the β-cyclodextrin derivatives exhibit very high binding potentials towards the anaesthetic drugs sevoflurane and halothane. Since the resulting inclusion compounds are highly soluble in water at temperatures ≤37 °C they are good candidates for new aqueous dosage forms which would avoid inhalation anaesthesia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90450
Triethylene glycol monomethyl ether, purum, ≥97.0% (GC)
C7H16O4