EMAIL THIS PAGE TO A FRIEND
Frontiers in bioscience (Elite edition)
Understanding the functional discrepancy of Pim-1 in cancer.
PMID 25553374
Abstract
The Pim-1 gene encodes for a proto-oncogenic serine/threonine protein kinase and is generally involved in cytokine signaling as well as in various signaling pathways regulating cell cycle and apoptosis. Pim-1 kinase plays a role in the development of various tumors mainly, prostate cancer, Burkitt's lymphoma, oral cancer and various other hematopoietic lymphomas. This review will focus on the importance and mechanisms of Pim-1 in prostate cancer and the potential clinical relevance of its various inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN1049 CompoZr® Knockout ZFN Kit, human PIM1 (NM_002648)