EMAIL THIS PAGE TO A FRIEND
BMC Biology
Dynamic response of RNA editing to temperature in Drosophila.
PMID 25555396
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1501
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®