EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Silk fibroin/gelatin electrospun nanofibrous dressing functionalized with astragaloside IV induces healing and anti-scar effects on burn wound.
PMID 25556053