EMAIL THIS PAGE TO A FRIEND
Glia
IL-17A activates ERK1/2 and enhances differentiation of oligodendrocyte progenitor cells.
PMID 25557204
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1255
Calcein Blue
C15H15NO7