EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research. Part B, Applied Biomaterials
Bioactive, nanostructured Si-substituted hydroxyapatite coatings on titanium prepared by pulsed laser deposition.
PMID 25557461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
450146
Calcium hydroxide, 99.995% trace metals basis
H2CaO2