EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
In vitro antifungal activity and in vivo antibiofilm activity of cerium nitrate against Candida species.
PMID 25558078
Abstract
The objective of this study was to clarify the antifungal properties of cerium, a lanthanide member, against Candida species. A comprehensive study with planktonic and sessile cells was performed. The ability of cerium nitrate (CN) to impair in vitro and in vivo biofilm formation was evaluated and its potential use in biofilm treatment was also evaluated. Forty-eight clinical isolates of different Candida species and the type strain ATCC 90028 were tested according to the protocol M27-A3. The MICs and minimum lethal concentrations were determined. A time-kill assay was performed and a cytometric kinetic study was performed using live/dead markers. Biofilm inhibition and biofilm susceptibility in the presence of cerium was evaluated by quantification of the biofilm metabolic activity and total biomass with XTT and crystal violet assays, respectively. CN in vivo efficacy as a coating for medical indwelling devices was evaluated for the first time for Candida parapsilosis, using a mouse subcutaneous foreign body model using polyurethane catheter segments. Scanning electron microscopy was used to assess biofilm architecture after CN treatment. The MICs for planktonic cells correlated with severe cellular metabolic activity impairment and membrane damage after 3 h of incubation. Moreover, CN efficiently prevented biofilm formation both in vitro and in vivo in segments of polyurethane catheters. At higher concentrations, it was also able to disorganize and almost eradicate preformed biofilms. Our results strongly suggest that CN application in the clinical setting might be effective in preventing the formation of biofilm-associated infections, namely through catheter coating and ultimately as an antimicrobial lock therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
461210
Cerium, chips
Ce
261041
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Ce
263001
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Ce
GF97217220
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Ce
GF49222582
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Ce
GF61818363
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF27295724
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF97378677
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Ce
GF36157394
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Ce
GF90687695
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ce
GF39030353
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Ce
GF04444095
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Ce
GF55512244
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Ce
GF02435554
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Ce
GF35961655
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ce
GF49695047
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Ce
202991
Cerium(III) nitrate hexahydrate, 99.999% trace metals basis
CeN3O9 · 6H2O
392219
Cerium(III) nitrate hexahydrate, 99.99% trace metals basis
CeN3O9 · 6H2O
238538
Cerium(III) nitrate hexahydrate, 99% trace metals basis
CeN3O9 · 6H2O