EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
Elastomeric core/shell nanofibrous cardiac patch as a biomimetic support for infarcted Porcine myocardium.
PMID 25559869
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29238
Activated Charcoal Norit®, Norit RBAA-3, rod
C
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
C
GF44361457
Carbon - Vitreous, foil, 200x200mm, thickness 1.0mm, glassy carbon
C
GF10355958
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
C
GF89520521
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
C