EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part C, Methods
Induction of Hepatic and Endothelial Differentiation by Perfusion in a Three-Dimensional Cell Culture Model of Human Fetal Liver.
PMID 25559936